Assay Details
Target Gene Details
Entrez Gene ID: | 15894 |
Gene Name: | intercellular adhesion molecule 1 |
Gene Aliases: |
CD54, Icam-1, Ly-47, MALA-2 |
Location: |
Chr.9:21015960-21028797 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Icam1 | NM_010493.2 | 4 | 816 | NP_034623.1 |
| AK149748.1 | 4 | 792 | BAE29059.1 | |
| AK149781.1 | 4 | 791 | BAE29079.1 | |
| AK149945.1 | 4 | 791 | ||
| AK150003.1 | 4 | 792 | ||
| AK150049.1 | 4 | 791 | BAE29266.1 | |
| AK150057.1 | 4 | 792 | BAE29272.1 | |
| AK150141.1 | 3 | 529 | BAE29336.1 | |
| AK151227.1 | 4 | 792 | BAE30219.1 | |
| AK151681.1 | 4 | 792 | ||
| AK152155.1 | 4 | 792 | BAE30990.1 | |
| AK152527.1 | 4 | 792 | BAE31286.1 | |
| AK152530.1 | 4 | 792 | BAE31287.1 | |
| AK152556.1 | 4 | 792 | BAE31310.1 | |
| AK156417.1 | 4 | 792 | ||
| AK168275.1 | 4 | 792 | BAE40221.1 | |
| AK171502.1 | 4 | 792 | BAE42493.1 | |
| AK171520.1 | 3 | 529 | BAE42504.1 | |
| AK172321.1 | 4 | 792 | BAE42944.1 | |
| BC008626.1 | 4 | 771 | AAH08626.1 | |
| CT010246.1 | 4 | 741 | CAJ18454.1 | |
| CT010302.1 | 4 | 741 | CAJ18510.1 | |
| M31585.1 | 4 | 799 | AAA37876.1 | |
| X16624.1 | 4 | 770 | CAA34621.1 | |
| X52264.1 | 4 | 763 | CAA36507.1 |

Genomic Map