Assay Details
Target Gene Details
Entrez Gene ID: | 15894 |
Gene Name: | intercellular adhesion molecule 1 |
Gene Aliases: |
CD54, Icam-1, Ly-47, MALA-2 |
Location: |
Chr.9:21015960-21028797 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Icam1 | NM_010493.2 | 7 | 1860 | NP_034623.1 |
AK149748.1 | 7 | 1836 | BAE29059.1 | |
AK149781.1 | 7 | 1835 | BAE29079.1 | |
AK149945.1 | 7 | 1835 | ||
AK150003.1 | 7 | 1836 | ||
AK150049.1 | 7 | 1835 | BAE29266.1 | |
AK150057.1 | 7 | 1836 | BAE29272.1 | |
AK151227.1 | 7 | 1836 | BAE30219.1 | |
AK151681.1 | 7 | 1836 | ||
AK152155.1 | 7 | 1836 | BAE30990.1 | |
AK152527.1 | 7 | 1836 | BAE31286.1 | |
AK152530.1 | 7 | 1836 | BAE31287.1 | |
AK152556.1 | 7 | 1836 | BAE31310.1 | |
AK156417.1 | 7 | 1834 | ||
AK168275.1 | 7 | 1836 | BAE40221.1 | |
AK171502.1 | 7 | 1836 | BAE42493.1 | |
AK171520.1 | 6 | 1573 | BAE42504.1 | |
AK172321.1 | 7 | 1836 | BAE42944.1 | |
BC008626.1 | 7 | 1815 | AAH08626.1 | |
M31585.1 | 7 | 1843 | AAA37876.1 | |
X16624.1 | 7 | 1814 | CAA34621.1 | |
X52264.1 | 7 | 1807 | CAA36507.1 |
