Assay Details
Target Gene Details
Entrez Gene ID: | 22599 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 20B |
Gene Aliases: |
AA986306, Sit1, Slc6a20, XT3, Xtrp3 |
Location: |
Chr.9:123593820-123632565 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a20b | NM_011731.3 | 1 | 340 | NP_035861.2 |
| AF075261.1 | 1 | 164 | AAC27756.1 | |
| AK143893.1 | 1 | 310 | ||
| AK144038.1 | 1 | 341 | BAE25668.1 | |
| AK163340.1 | 1 | 103 | ||
| AK164706.1 | 1 | 185 | BAE37885.1 | |
| AK164713.1 | 1 | 175 | ||
| BC013484.1 | 1 | 251 | AAH13484.1 |
More Information
Set Membership: |
|

Genomic Map