Assay Details
Target Gene Details
Entrez Gene ID: | 235631 |
Gene Name: | protease, serine 50 |
Gene Aliases: |
79H19C, Tsp50 |
Location: |
Chr.9:110857899-110864629 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss50 | NM_146227.4 | 1 | 411 | NP_666339.2 |
| AF481144.1 | 1 | 397 | AAM89223.1 | |
| AK045168.1 | 1 | 412 | BAC32245.1 | |
| BC150686.1 | 1 | 410 |
More Information
Set Membership: |
|

Genomic Map