Assay Details
Target Gene Details
Entrez Gene ID: | 72472 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 10 |
Gene Aliases: |
2610103N14Rik, 9830169E08, Mct10, PRO0813, TAT1 |
Location: |
Chr.10:40033532-40142264 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc16a10 | NM_001114332.1 | 3 | 852 | NP_001107804.1 |
| NM_028247.4 | 3 | 852 | NP_082523.1 | |
| AK036743.1 | 3 | 853 | ||
| AK050229.1 | 3 | 855 | BAC34135.1 | |
| AK144135.1 | 3 | 855 | ||
| AK151290.1 | 3 | 853 | BAE30275.1 | |
| AK151636.1 | 3 | 854 | ||
| AK151704.1 | 3 | 857 | BAE30627.1 | |
| AK157355.1 | 3 | 841 | BAE34061.1 | |
| AK165240.1 | 3 | 853 | BAE38097.1 | |
| AK165579.1 | 3 | 855 | BAE38270.1 | |
| BC052877.1 | 3 | 333 | AAH52877.1 |
More Information
Set Membership: |
|

Genomic Map