Assay Details
Target Gene Details
Entrez Gene ID: | 72472 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 10 |
Gene Aliases: |
2610103N14Rik, 9830169E08, Mct10, PRO0813, TAT1 |
Location: |
Chr.10:40033532-40142264 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc16a10 | NM_001114332.1 | 6 | 2822 | NP_001107804.1 |
| AK149461.1 | 1 | 1100 | BAE28894.1 | |
| AK165240.1 | 6 | 2823 | BAE38097.1 |
More Information
Set Membership: |
|

Genomic Map