Assay Details
Target Gene Details
Entrez Gene ID: | 69678 |
Gene Name: | ataxia, cerebellar, Cayman type homolog (human), opposite strand |
Gene Aliases: |
2310050B05Rik |
Location: |
Chr.10:81194691-81208370 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atcayos | NR_015477.1 | |||
| AK084504.1 |
Target Gene Details
Entrez Gene ID: | 69564 |
Gene Name: | nicotinamide riboside kinase 2 |
Gene Aliases: |
2310015C21Rik, Itgb1bp3, Mibp |
Location: |
Chr.10:81198170-81202212 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nmrk2 | NM_027120.2 | 7 | 989 | NP_081396.1 |
| AK137483.1 | 7 | 991 | BAE23375.1 |
More Information
Set Membership: |
|

Genomic Map