Assay Details
Target Gene Details
Entrez Gene ID: | 68632 |
Gene Name: | myc target 1 |
Gene Aliases: |
1110020B04Rik, AI225941, AI642973, Mtmc1 |
Location: |
Chr.10:5593728-5606789 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myct1 | NM_026793.2 | 2 | 1916 | NP_081069.1 |
AK030706.1 | 2 | 1841 | BAE20454.1 | |
AK054222.1 | 2 | 1815 | BAC35696.1 | |
AK142963.1 | 2 | 1849 | BAE25233.1 |
