Assay Details
Target Gene Details
Entrez Gene ID: | 56516 |
Gene Name: | RNA binding motif, single stranded interacting protein 2 |
Gene Aliases: |
2610315E04Rik, Scr3 |
Location: |
Chr.10:128129469-128180297 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rbms2 | NM_001039080.1 | 14 | 3448 | NP_001034169.1 |
NM_019711.2 | 13 | 3265 | NP_062685.2 |
