Assay Details
Target Gene Details
Entrez Gene ID: | 13688 |
Gene Name: | eukaryotic translation initiation factor 4E binding protein 2 |
Gene Aliases: |
2810011I19Rik, 4E-BP2, AA792569, BC010348, PHAS-II |
Location: |
Chr.10:61432497-61452669 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4ebp2 | NM_010124.2 | 1 | 224 | NP_034254.1 |
| AK133912.1 | 1 | 112 | BAE21923.1 | |
| AK177357.1 | ||||
| AK194349.1 | ||||
| BC015082.1 | 2 | 205 | AAH15082.1 | |
| BC119539.2 | 2 | 188 | ||
| U75530.1 | 1 | 224 | AAC52899.1 |
More Information
Set Membership: |
|

Genomic Map