Assay Details
Target Gene Details
Entrez Gene ID: | 237459 |
Gene Name: | cyclin-dependent kinase 17 |
Gene Aliases: |
6430598J10Rik, Pctk2 |
Location: |
Chr.10:93160876-93250688 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk17 |
Target Gene Details
Entrez Gene ID: | 13713 |
Gene Name: | ELK3, member of ETS oncogene family |
Gene Aliases: |
D430049E23Rik, Erp, Etrp, Net, Sap-2 |
Location: |
Chr.10:93247415-93311167 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elk3 | NM_001282967.1 | 4 | 2096 | NP_001269896.1 |
| NM_013508.2 | 5 | 2897 | NP_038536.2 | |
| AK052546.1 | 5 | 2574 | BAC35034.1 |
More Information
Set Membership: |
|

Genomic Map