Assay Details
Target Gene Details
Entrez Gene ID: | 66399 |
Gene Name: | Ts translation elongation factor, mitochondrial |
Gene Aliases: |
2310050B20Rik, 9430024O13Rik, EF-TS, EF-Tsmt |
Location: |
Chr.10:127022332-127030814 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tsfm | NM_025537.3 | 6 | 980 | NP_079813.1 |
| AK009898.1 | 6 | 1001 | ||
| AK012240.1 | 6 | 971 | BAB28113.1 | |
| AK018591.1 | 6 | 969 | ||
| AK169164.1 | 6 | 971 | BAE40943.1 | |
| AK171656.1 | 6 | 977 | BAE42591.1 | |
| AK172112.1 | 6 | 977 | BAE42833.1 | |
| AK210440.1 | 1 | 70 | ||
| BC034286.1 | 6 | 959 | AAH34286.1 | |
| BC051508.1 | 3 | 561 | AAH51508.1 | |
| BC057904.1 | 6 | 980 | AAH57904.1 |
More Information
Set Membership: |
|

Genomic Map