Assay Details
Target Gene Details
Entrez Gene ID: | 13853 |
Gene Name: | epilepsy, progressive myoclonic epilepsy, type 2 gene alpha |
Gene Aliases: |
TG-B, Tg(TcraK,TcrbK)TG-BFlv |
Location: |
Chr.10:11342690-11457477 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epm2a | NM_010146.2 | 1 | 267 | NP_034276.2 |
AF124044.1 | 1 | 308 | AAD26336.1 | |
AK041609.1 | 1 | 214 | BAC31004.1 | |
AK155519.1 | 1 | 281 | ||
BY730431.1 | 1 | 268 |
