Assay Details
Target Gene Details
Entrez Gene ID: | 320642 |
Gene Name: | RIKEN cDNA A630066F11 gene |
Gene Aliases: |
- |
Location: |
Chr.10:7663371-7664623 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A630066F11Rik | NR_030698.1 | 1 | 43 | |
| AK039358.1 | ||||
| AK042171.1 | ||||
| AK083146.1 | 1 | 44 |
Target Gene Details
Entrez Gene ID: | 18537 |
Gene Name: | protein-L-isoaspartate (D-aspartate) O-methyltransferase 1 |
Gene Aliases: |
C79501, PIMT |
Location: |
Chr.10:7629380-7680973 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcmt1 | NM_008786.2 | 1 | 172 | NP_032812.2 |
| AK148789.1 | 1 | 197 |
More Information
Set Membership: |
|

Genomic Map