Assay Details
Target Gene Details
Entrez Gene ID: | 68265 |
Gene Name: | IQ motif containing F3 |
Gene Aliases: |
1700012F17Rik, AV040440 |
Location: |
Chr.9:106504666-106561631 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqcf3 |
Target Gene Details
Entrez Gene ID: | 75470 |
Gene Name: | IQ motif containing F5 |
Gene Aliases: |
1700007L12Rik |
Location: |
Chr.9:106514573-106516003 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqcf5 | NM_029300.1 | 2 | 390 | NP_083576.1 |
| AK005734.1 | 2 | 391 | BAB24212.1 | |
| BC125546.1 | 2 | 349 |

Genomic Map