Assay Details
Target Gene Details
Entrez Gene ID: | 66712 |
Gene Name: | sperm equatorial segment protein 1 |
Gene Aliases: |
4921508E09Rik, AI428149 |
Location: |
Chr.9:62270729-62282179 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spesp1 | NM_025721.2 | 2 | 1594 | NP_079997.1 |
AK014843.1 | 2 | 1595 | BAB29577.1 |
