Assay Details
Target Gene Details
Entrez Gene ID: | 50493 |
Gene Name: | thioredoxin reductase 1 |
Gene Aliases: |
TR, TR1, TrxR1 |
Location: |
Chr.10:82833951-82897724 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Txnrd1 | NM_001042513.1 | NP_001035978.1 | ||
| NM_001042514.1 | NP_001035979.1 | |||
| NM_001042523.1 | 2 | 307 | NP_001035988.1 | |
| NM_015762.2 | NP_056577.2 | |||
| AB027565.1 | BAA86985.2 | |||
| AF333036.1 | 2 | 307 | AAK01140.1 | |
| AK011902.1 | BAB27905.1 | |||
| AK087645.1 | 1 | 813 | ||
| AK146125.1 | BAE26918.1 | |||
| AK149625.1 | BAE28994.1 | |||
| AK151927.1 | ||||
| AK168356.1 | BAE40292.1 | |||
| AK172383.1 | ||||
| AK177272.1 | ||||
| AK180432.1 | ||||
| AK184063.1 | ||||
| AK185936.1 | ||||
| AK186760.1 | ||||
| AK186863.1 | ||||
| AK186897.1 | ||||
| AK194169.1 | ||||
| AK210892.1 | ||||
| BC037643.1 | AAH37643.1 | |||
| CJ136679.1 | ||||
| CX562215.1 |
More Information
Set Membership: |
|

Genomic Map