Assay Details
Target Gene Details
Entrez Gene ID: | 12461 |
Gene Name: | chaperonin containing Tcp1, subunit 2 (beta) |
Gene Aliases: |
Cctb |
Location: |
Chr.10:117050997-117063814 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cct2 | NM_007636.2 | 16 | 1819 | NP_031662.2 |
| AA023539.1 | 1 | 151 | ||
| AB041570.1 | 16 | 1650 | BAA95054.1 | |
| AK012295.1 | 16 | 1779 | ||
| AK075581.1 | 16 | 1792 | BAC35834.1 | |
| AK088145.1 | 16 | 1781 | ||
| AK145406.1 | 16 | 1820 | BAE26419.1 | |
| AK166891.1 | 16 | 1792 | BAE39096.1 | |
| BC007470.1 | 16 | 1783 | AAH07470.1 | |
| BC026918.1 | 16 | 1785 | AAH26918.1 | |
| Z31553.1 | 16 | 1770 | CAA83428.1 |
More Information
Set Membership: |
|

Genomic Map