Assay Details
Target Gene Details
Entrez Gene ID: | 53599 |
Gene Name: | CD164 antigen |
Gene Aliases: |
A115, A24, AA589639, AW540279, MGC-24, MSSP |
Location: |
Chr.10:41519500-41531042 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd164 | NM_016898.2 | 6 | 2429 | NP_058594.1 |
AB028895.1 | 6 | 2429 | BAA78909.1 | |
AF299345.1 | 6 | 2394 | AAG53910.1 | |
AK002792.1 | 1 | 1875 | BAB22362.2 | |
AK003242.1 | 1 | 152 | ||
AK145131.1 | 6 | 2488 | BAE26251.1 | |
AK169275.1 | 6 | 2469 | BAE41034.1 | |
AK207902.1 | ||||
AK217133.1 | ||||
BC005414.1 | 6 | 2417 | AAH05414.1 |
