Assay Details
Target Gene Details
Entrez Gene ID: | 215819 |
Gene Name: | NHS-like 1 |
Gene Aliases: |
5730409E15Rik, A630035H13Rik, A730096F01, BC013565, D10Bwg0940e, mKIAA1357 |
Location: |
Chr.10:18318558-18533892 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nhsl1 | NM_001163592.1 | 6 | 2418 | NP_001157064.1 |
| NM_173390.3 | 5 | 2941 | NP_775566.3 |

Genomic Map