Assay Details
Target Gene Details
Entrez Gene ID: | 56441 |
Gene Name: | N-acetyltransferase 6 |
Gene Aliases: |
AI225910, Fus2 |
Location: |
Chr.9:107580170-107584048 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nat6 | NM_019750.3 | 1 | 512 | NP_062724.1 |
| BC026545.1 | 1 | 537 | AAH26545.1 |
More Information
Set Membership: |
|

Genomic Map