Assay Details
Target Gene Details
Entrez Gene ID: | 71713 |
Gene Name: | cell division cycle 40 |
Gene Aliases: |
1200003H23Rik, EHB3, PRP17 |
Location: |
Chr.10:40831621-40883143 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc40 | NM_027879.2 | 1 | 136 | NP_082155.1 |
| AK004569.1 | 1 | 103 | BAB23380.1 | |
| AK040680.1 | 1 | 85 | ||
| AK084140.1 | 1 | 134 | ||
| AK135411.1 | 1 | 103 | BAE22524.1 | |
| BC061220.1 | 1 | 111 | AAH61220.1 | |
| BC147503.1 | 1 | 136 | ||
| BC158078.1 | 1 | 136 |
More Information
Set Membership: |
|

Genomic Map