Assay Details
Target Gene Details
Entrez Gene ID: | 268354 |
Gene Name: | family with sequence similarity 19, member A2 |
Gene Aliases: |
6330575M02, FAM19A2 TAFA-2, TAFA2, Tafa-2 |
Location: |
Chr.10:123264076-123741204 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam19a2 | NM_001252387.1 | 1 | 237 | NP_001239316.1 |
| NM_182807.3 | 1 | 237 | NP_877959.2 | |
| NR_045514.1 | 1 | 237 | ||
| AK139403.1 | 1 | 317 | BAE23995.1 | |
| AK140763.1 | 1 | 200 | BAE24471.1 | |
| BC027082.1 | 1 | 123 | AAH27082.1 | |
| CD350359.1 | 1 | 237 |
More Information
Set Membership: |
|

Genomic Map