Assay Details
Target Gene Details
Entrez Gene ID: | 268354 |
Gene Name: | family with sequence similarity 19, member A2 |
Gene Aliases: |
6330575M02, FAM19A2 TAFA-2, TAFA2, Tafa-2 |
Location: |
Chr.10:123264076-123741204 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam19a2 | NM_001252387.1 | 5 | 2196 | NP_001239316.1 |
| NM_182807.3 | 6 | 2408 | NP_877959.2 | |
| NR_045514.1 | 6 | 2442 | ||
| AK032077.1 | 3 | 512 | ||
| AK032483.1 | 5 | 1278 | ||
| AK139403.1 | 6 | 2526 | BAE23995.1 | |
| AK140763.1 | 6 | 2405 | BAE24471.1 | |
| AK164201.1 | 5 | 1288 | BAE37679.1 | |
| AY325121.1 | AAP92413.1 | |||
| BC027082.1 | 6 | 2294 | AAH27082.1 |
More Information
Set Membership: |
|

Genomic Map