Assay Details
Target Gene Details
Entrez Gene ID: | 237403 |
Gene Name: | leucine rich repeat and Ig domain containing 3 |
Gene Aliases: |
AI851712, LERN2 |
Location: |
Chr.10:80832801-80844039 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lingo3 | NM_001013758.2 | 2 | 2887 | NP_001013780.2 |
| BC072620.1 | 2 | 2878 | AAH72620.1 |
More Information
Set Membership: |
|

Genomic Map