Assay Details
Target Gene Details
Entrez Gene ID: | 215819 |
Gene Name: | NHS-like 1 |
Gene Aliases: |
5730409E15Rik, A630035H13Rik, A730096F01, BC013565, D10Bwg0940e, mKIAA1357 |
Location: |
Chr.10:18318558-18533892 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nhsl1 | NM_001163592.1 | 8 | 5037 | NP_001157064.1 |
| NM_173390.3 | 7 | 5560 | NP_775566.3 | |
| AK043447.1 | 3 | 1486 | BAC31551.1 | |
| AK134837.1 | 3 | 1508 | BAE22306.1 | |
| AK220386.1 | 3 | 1605 | BAD90251.1 | |
| BC013565.1 | 3 | 1159 | AAH13565.2 | |
| BC061949.1 | 3 | 1140 | AAH61949.1 |

Genomic Map