Assay Details
Target Gene Details
Entrez Gene ID: | 215789 |
Gene Name: | phosphatase and actin regulator 2 |
Gene Aliases: |
AV158170, BC012871 |
Location: |
Chr.10:13207717-13474521 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phactr2 | NM_001033257.4 | 5 | 958 | NP_001028429.1 |
| NM_001195065.1 | 6 | 1214 | NP_001181994.1 | |
| NM_001195066.1 | 6 | 1149 | NP_001181995.1 | |
| NM_001195096.1 | 6 | 970 | NP_001182025.1 | |
| AK139466.1 | 6 | 1150 | BAE24022.1 | |
| AK142930.1 | 5 | 959 | BAE25225.1 | |
| BC012871.1 | 6 | 1238 | AAH12871.1 | |
| BC130264.1 | 6 | 966 |
More Information
Set Membership: |
|

Genomic Map