Assay Details
Target Gene Details
Entrez Gene ID: | 68031 |
Gene Name: | ring finger protein 146 |
Gene Aliases: |
2610509H23Rik, Iduna |
Location: |
Chr.10:29344176-29362442 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf146 | NM_001110196.1 | 1 | 160 | NP_001103666.1 |
| NM_001110197.1 | 1 | 160 | NP_001103667.1 | |
| NM_001110198.1 | 1 | 160 | NP_001103668.1 | |
| NM_026518.4 | 1 | 160 | NP_080794.2 | |
| BC050795.1 | 1 | 160 | AAH50795.1 |

Genomic Map