Assay Details
Target Gene Details
Entrez Gene ID: | 11537 |
Gene Name: | complement factor D (adipsin) |
Gene Aliases: |
Adn, DF |
Location: |
Chr.10:79890853-79892660 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfd | NM_001291915.1 | 4 | 600 | NP_001278844.1 |
| NM_013459.3 | 4 | 603 | NP_038487.1 | |
| AK143817.1 | 4 | 547 | BAE25550.1 | |
| BC138778.1 | 4 | 603 | ||
| BC138779.1 | 4 | 603 | ||
| BC145412.1 | 4 | 600 | ||
| BE136302.1 | 2 | 324 | ||
| DV051476.1 | 4 | 548 | ||
| M11768.1 | 4 | 544 | AAA40486.1 |
More Information
Set Membership: |
|

Genomic Map