Assay Details
Target Gene Details
Entrez Gene ID: | 94221 |
Gene Name: | golgi associated PDZ and coiled-coil motif containing |
Gene Aliases: |
2210402P09Rik, AI844555, CAL, FIG, GOPC1, PIST |
Location: |
Chr.10:52337024-52382124 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gopc | NM_001199272.1 | 9 | 3348 | NP_001186201.1 |
| NM_053187.3 | 8 | 3324 | NP_444417.2 | |
| AK008806.1 | 1 | 509 | ||
| AK009536.1 | ||||
| AK030427.1 | 9 | 2944 | BAC26958.1 | |
| BC051171.1 | 8 | 3275 | AAH51171.1 |
More Information
Set Membership: |
|

Genomic Map