Assay Details
Target Gene Details
Entrez Gene ID: | 237362 |
Gene Name: | neuropeptide FF receptor 1 |
Gene Aliases: |
Gm236, Gpr147, NPFF1, OT7T022 |
Location: |
Chr.10:61595484-61628762 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npffr1 | NM_001177511.1 | 3 | 1009 | NP_001170982.1 |
More Information
Set Membership: |
|

Genomic Map