Assay Details
Target Gene Details
Entrez Gene ID: | 238161 |
Gene Name: | A kinase (PRKA) anchor protein 6 |
Gene Aliases: |
AI482140, Akapalpha, Akapbeta, PRKA |
Location: |
Chr.12:52699016-53155568 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akap6 | NM_198111.2 | 13 | 5360 | NP_932779.2 |
| AK129115.1 | 1 | 819 | BAC97925.1 | |
| AK147248.1 | 13 | 5361 | BAE27795.1 | |
| AK147371.1 | 13 | 5361 | BAE27869.1 | |
| AK162929.1 | 2 | 1755 | BAE37120.1 | |
| BC057114.1 | AAH57114.1 | |||
| DQ233645.1 | 13 | 5416 | ABB60211.1 | |
| DQ233646.1 | 12 | 5210 | ABB60212.1 |

Genomic Map