Assay Details
Target Gene Details
Entrez Gene ID: | 238161 |
Gene Name: | A kinase (PRKA) anchor protein 6 |
Gene Aliases: |
AI482140, Akapalpha, Akapbeta, PRKA |
Location: |
Chr.12:52699016-53155568 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Akap6 | NM_198111.2 | 3 | 502 | NP_932779.2 |
AK147248.1 | 3 | 503 | BAE27795.1 | |
AK147371.1 | 3 | 503 | BAE27869.1 | |
BC057114.1 | 2 | 452 | AAH57114.1 | |
DQ233645.1 | 3 | 555 | ABB60211.1 | |
DQ233646.1 | 2 | 349 | ABB60212.1 |
