Assay Details
Target Gene Details
Entrez Gene ID: | 67131 |
Gene Name: | acyl-Coenzyme A binding domain containing 4 |
Gene Aliases: |
2010009P05Rik, 2010015A21Rik, AI849317 |
Location: |
Chr.11:103101688-103112199 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acbd4 | NM_025988.2 | 2 | 172 | NP_080264.1 |
| AK079604.1 | 2 | 167 | BAC37699.1 | |
| BC048371.1 | 1 | 86 | AAH48371.1 | |
| DN173459.1 | 2 | 129 |
More Information
Set Membership: |
|

Genomic Map