Assay Details
Target Gene Details
Entrez Gene ID: | 104112 |
Gene Name: | ATP citrate lyase |
Gene Aliases: |
A730098H14Rik, AW538652 |
Location: |
Chr.11:100476352-100528000 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acly | NM_001199296.1 | 29 | 4259 | NP_001186225.1 |
| NM_134037.3 | 28 | 4229 | NP_598798.1 | |
| AK043434.1 | 12 | 2155 | ||
| AK132755.1 | 29 | 4150 | BAE21336.1 | |
| AK135528.1 | 29 | 4127 | ||
| AK136812.1 | 12 | 2067 | BAE23135.1 | |
| AK169703.1 | 28 | 4077 | BAE41315.1 | |
| BC005533.1 | 6 | 1312 | AAH05533.1 | |
| BC021502.1 | 15 | 2505 | AAH21502.1 | |
| BC056378.1 | 28 | 4130 | AAH56378.1 | |
| BC065805.1 | 4 | 1276 | AAH65805.1 | |
| BE623317.1 | 1 | 180 |
More Information
Set Membership: |
|

Genomic Map