Assay Details
Target Gene Details
Entrez Gene ID: | 104776 |
Gene Name: | aldehyde dehydrogenase family 6, subfamily A1 |
Gene Aliases: |
1110038I05Rik, AI314632, Mmsdh |
Location: |
Chr.12:84430717-84450950 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh6a1 | NM_134042.2 | 6 | 593 | NP_598803.1 |
| AF297860.1 | 6 | 593 | AAG44988.1 | |
| AK033587.1 | 6 | 620 | BAC28375.1 | |
| AK082790.1 | 6 | 647 | ||
| AK128924.1 | 6 | 625 | ||
| AK147146.1 | 6 | 647 | BAE27715.1 | |
| AK155814.1 | 6 | 644 | BAE33445.1 | |
| AK164366.1 | 5 | 2193 | ||
| AK169915.1 | 6 | 633 | BAE41455.1 | |
| AK170305.1 | 6 | 646 | BAE41702.1 | |
| AK171581.1 | 6 | 647 | BAE42539.1 | |
| AK171896.1 | 6 | 636 | BAE42726.1 | |
| AK183301.1 | 2 | 231 | ||
| AK197238.1 | 2 | 198 | ||
| AK212397.1 | 2 | 207 | ||
| AK217579.1 | 2 | 194 | ||
| AK219365.1 | 2 | 214 | ||
| BC031148.1 | 6 | 718 | AAH31148.1 | |
| BC033440.1 | 5 | 616 | AAH33440.1 |
Target Gene Details
Entrez Gene ID: | 72873 |
Gene Name: | coiled-coil domain containing 176 |
Gene Aliases: |
2900006K08Rik, AI427784 |
Location: |
Chr.12:84409068-84441763 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc176 |
More Information
Set Membership: |
|

Genomic Map