Assay Details
Target Gene Details
Entrez Gene ID: | 104184 |
Gene Name: | bleomycin hydrolase |
Gene Aliases: |
AI035728, Bh, Bmh |
Location: |
Chr.11:76945656-76987389 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Blmh | NM_178645.4 | 12 | 2191 | NP_848760.1 |
| AI035728.1 | 1 | 157 | ||
| AK035228.1 | 12 | 2065 | BAC28989.1 | |
| AK049461.1 | 12 | 2104 | ||
| AK051441.1 | 12 | 2090 | BAC34639.1 | |
| AK150438.1 | 12 | 2090 | BAE29560.1 | |
| AK150896.1 | 12 | 2071 | ||
| AK167325.1 | 12 | 2071 | BAE39427.1 | |
| AK180523.1 | 1 | 375 | ||
| BC027037.1 | 12 | 2047 | AAH27037.1 | |
| BC027403.1 | 12 | 2101 | AAH27403.1 | |
| BY643607.1 | 1 | 237 |
More Information
Set Membership: |
|

Genomic Map