Assay Details
Target Gene Details
Entrez Gene ID: | 65257 |
Gene Name: | ankyrin repeat and SOCS box-containing 3 |
Gene Aliases: |
2400011J03Rik, Asb-3 |
Location: |
Chr.11:30954194-31102704 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb3 | NM_023906.3 | NP_076395.2 | ||
| AK010315.1 | BAB26847.1 | |||
| AK028489.1 | ||||
| AK038086.1 | ||||
| AK038897.1 | ||||
| AK143580.1 | BAE25448.1 | |||
| BB618690.1 | ||||
| BC023086.1 | AAH23086.1 |
Target Gene Details
Entrez Gene ID: | 68044 |
Gene Name: | ChaC, cation transport regulator 2 |
Gene Aliases: |
- |
Location: |
Chr.11:30976707-30986365 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chac2 | NM_001290667.1 | 2 | 605 | NP_001277596.1 |
| NM_026527.3 | 3 | 641 | NP_080803.1 | |
| AK007629.1 | 3 | 565 | ||
| AK010914.1 | 3 | 611 | BAB27264.1 | |
| AK021391.1 | 2 | 575 | BAB32394.1 | |
| AK078993.1 | 3 | 627 | BAC37498.1 | |
| BC025100.1 | 3 | 555 | AAH25100.1 |
More Information
Set Membership: |
|

Genomic Map