Assay Details
Target Gene Details
Entrez Gene ID: | 276919 |
Gene Name: | gem (nuclear organelle) associated protein 4 |
Gene Aliases: |
4932415L08Rik, AU015359 |
Location: |
Chr.11:76210571-76217572 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gemin4 | NM_177367.3 | 2 | 653 | NP_796341.2 |
| AK133821.1 | 2 | 745 | ||
| AK141078.1 | 2 | 632 | BAE24559.1 | |
| AK154061.1 | 2 | 634 | BAE32348.1 | |
| AK165864.1 | 2 | 597 | ||
| BC031425.1 | 2 | 593 | AAH31425.1 | |
| BC062157.1 | 2 | 601 | AAH62157.1 |

Genomic Map