Assay Details
Target Gene Details
Entrez Gene ID: | 24083 |
Gene Name: | N-acetyltransferase domain containing 1 |
Gene Aliases: |
AI256713, F3-2, Gm16515, Gtlf3b |
Location: |
Chr.11:60902246-60913792 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Natd1 | NM_025294.5 | 3 | 2225 | NP_079570.1 |
| AK004720.2 | 3 | 2224 | BAB23503.1 | |
| AK080453.1 | 3 | 2225 | BAC37922.1 | |
| AK089179.1 | 3 | 2224 | BAC40780.1 | |
| AK179282.1 | 1 | 274 | ||
| AK182237.1 | 1 | 274 | ||
| AK207502.1 | 1 | 257 | ||
| AK215610.1 | 1 | 268 | ||
| BC034332.1 | 3 | 2181 | AAH34332.1 | |
| BC038816.1 | 3 | 2223 | AAH38816.1 |

Genomic Map