Assay Details
Target Gene Details
Entrez Gene ID: | 16661 |
Gene Name: | keratin 10 |
Gene Aliases: |
D130054E02Rik, K10, K1C1, Krt-1.10, Krt1-10 |
Location: |
Chr.11:99385254-99389699 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Krt10 | NM_010660.2 | 1 | 110 | NP_034790.2 |
AK014360.1 | 1 | 96 | BAB29296.1 | |
AK076508.1 | 1 | 108 | BAC36371.1 | |
AK081402.1 | 1 | 110 | BAC38210.1 | |
AK081914.1 | 1 | 107 | BAC38369.1 | |
BB648004.1 | 1 | 111 | ||
V00830.1 | 1 | 105 | CAA24214.1 |
