Assay Details
Target Gene Details
Entrez Gene ID: | 20850 |
Gene Name: | signal transducer and activator of transcription 5A |
Gene Aliases: |
AA959963, STAT5 |
Location: |
Chr.11:100859180-100885169 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stat5a | NM_001164062.1 | 18 | 2940 | NP_001157534.1 |
| NM_011488.3 | 20 | 3222 | NP_035618.1 | |
| AK134012.1 | 7 | 1341 | BAE21979.1 | |
| AK154070.1 | 19 | 2957 | BAE32355.1 | |
| BC013274.1 | 20 | 3181 | AAH13274.1 | |
| U21103.1 | 20 | 3070 | AAA80590.1 |
More Information
Set Membership: |
|

Genomic Map