Assay Details
Target Gene Details
Entrez Gene ID: | 20868 |
Gene Name: | serine/threonine kinase 10 |
Gene Aliases: |
Gek1, Lok, mKIAA4026 |
Location: |
Chr.11:32533266-32624595 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stk10 | NM_009288.2 | 19 | 3263 | NP_033314.2 |
AK148118.1 | 19 | 3203 | BAE28356.1 | |
AK220448.1 | 19 | 3185 | BAD90480.1 | |
BC128363.1 | 19 | 3149 | ||
D89728.1 | 19 | 3224 | BAA24073.1 |
