Assay Details
Target Gene Details
Entrez Gene ID: | 74319 |
Gene Name: | methyltransferase like 23 |
Gene Aliases: |
1110005A03Rik, 1500035B17Rik, 4933424L15Rik |
Location: |
Chr.11:116843515-116849740 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mettl23 | NM_028865.3 | 1 | 43 | NP_083141.3 |
AK168267.1 | ||||
BC026936.1 | AAH26936.1 | |||
BY122329.1 | 1 | 43 |
