Assay Details
Target Gene Details
Entrez Gene ID: | 16667 |
Gene Name: | keratin 17 |
Gene Aliases: |
K17, Krt1-17 |
Location: |
Chr.11:100256217-100260989 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Krt17 | NM_010663.2 | 1 | 160 | NP_034793.1 |
AB013608.1 | 1 | 161 | BAA34229.1 | |
BC132454.1 | 1 | 195 | ||
BC132456.1 | 1 | 195 |
