Assay Details
Target Gene Details
Entrez Gene ID: | 71966 |
Gene Name: | NFKB inhibitor interacting Ras-like protein 2 |
Gene Aliases: |
2410003M04Rik, 4930527H08Rik, D630018G21Rik, KBRAS2 |
Location: |
Chr.11:100622955-100627602 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nkiras2 | NM_028024.2 | 4 | 675 | NP_082300.1 |
AK010371.1 | 4 | 675 | BAB26889.1 | |
AK052645.1 | 4 | 675 | BAC35078.1 | |
AK083475.1 | 4 | 718 | BAC38929.1 | |
AK149696.1 | 4 | 819 | BAE29033.1 | |
AK169878.1 | 4 | 700 | BAE41430.1 | |
AK179600.1 | ||||
AK208908.1 | ||||
BC013469.1 | 3 | 628 | AAH13469.1 | |
BC024398.1 | 3 | 618 | AAH24398.1 |
Target Gene Details
Entrez Gene ID: | 278304 |
Gene Name: | zinc finger protein 385C |
Gene Aliases: |
A930006D11Rik, Znf385c |
Location: |
Chr.11:100626219-100692659 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp385c | NM_177790.4 | 8 | 4045 | NP_808458.2 |
