Assay Details
Target Gene Details
Entrez Gene ID: | 80879 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 3 |
Gene Aliases: |
Mct3, Mct4 |
Location: |
Chr.11:120948484-120959000 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc16a3 | NM_001038653.1 | 5 | 1205 | NP_001033742.1 |
| NM_001038654.1 | 4 | 1110 | NP_001033743.1 | |
| NM_030696.3 | 5 | 1217 | NP_109621.1 | |
| AF178954.1 | 3 | 1005 | AAG24271.1 | |
| AK141381.1 | 4 | 1107 | ||
| AK149707.1 | 5 | 1208 | BAE29040.1 | |
| AK149983.1 | 5 | 1208 | BAE29212.1 | |
| AK165596.1 | 4 | 1053 | ||
| AK169917.1 | 5 | 1167 | BAE41457.1 | |
| AK171260.1 | 5 | 1193 | BAE42351.1 | |
| BC046525.1 | 5 | 1217 | AAH46525.1 |
More Information
Set Membership: |
|

Genomic Map