Assay Details
Target Gene Details
Entrez Gene ID: | 104318 |
Gene Name: | casein kinase 1, delta |
Gene Aliases: |
1200006A05Rik, AA409348, D930010H05Rik |
Location: |
Chr.11:120958010-120991333 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk1d | AK140317.1 | 9 | 1729 | BAE24329.1 |
Target Gene Details
Entrez Gene ID: | 80879 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 3 |
Gene Aliases: |
Mct3, Mct4 |
Location: |
Chr.11:120948484-120959000 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc16a3 | NM_001038653.1 | 6 | 2293 | NP_001033742.1 |
| NM_001038654.1 | 5 | 2198 | NP_001033743.1 | |
| NM_030696.3 | 6 | 2305 | NP_109621.1 | |
| AK165596.1 | 5 | 2142 |
More Information
Set Membership: |
|

Genomic Map