Assay Details
Target Gene Details
Entrez Gene ID: | 217316 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 5 |
Gene Aliases: |
A130015N09Rik, MCT 5, MCT 6, MCT5 |
Location: |
Chr.11:115462250-115474398 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc16a5 | NM_001080934.1 | 3 | 875 | NP_001074403.1 |
| AK037416.1 | 2 | 987 | ||
| AK079757.1 |
More Information
Set Membership: |
|

Genomic Map