Assay Details
Target Gene Details
Entrez Gene ID: | 68107 |
Gene Name: | cyclin N-terminal domain containing 1 |
Gene Aliases: |
1700051C09Rik, 4921513J16Rik, 5430417M23Rik, Cntd |
Location: |
Chr.11:101279203-101288701 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cntd1 | NM_026562.2 | 1 | 53 | NP_080838.1 |
AK006754.1 | BAB24728.1 | |||
AK007245.1 | 1 | 54 | BAB24912.1 | |
AK049116.1 | 1 | 161 | BAC33551.1 | |
BC006866.1 | 1 | 112 | AAH06866.1 |
